Thermo Scientific Chemicals

Zinc propionate, 97%, Thermo Scientific Chemicals

Catalog number: B23636.30
250 g, Each
Thermo Scientific Chemicals

Zinc propionate, 97%, Thermo Scientific Chemicals

Catalog number: B23636.30
250 g, Each
Quantity
Catalog number: B23636.30
also known as B23636-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-28-8
Form
Powder
Appearance (Color)
White
Complexometric Titration
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

Solubility
Soluble in alcohol, ether, chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.

Notes
Hygroscopic, protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text