Thermo Scientific Chemicals

Thiophene-3-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23637.06
5 g, Each
Thermo Scientific Chemicals

Thiophene-3-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23637.06
5 g, Each
Quantity
Catalog number: B23637.06
also known as B23637-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6165-69-1
IUPAC Name
(thiophen-3-yl)boronic acid
Molecular Formula
C4H5BO2S
InChI Key
QNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILES
OB(O)C1=CSC=C1
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text