2-Hydroxy-3-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-3-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxy-3-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23658.03
also known as B23658-03
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
2-Hydroxy-3-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB23658.03
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS5274-70-4
IUPAC Name2-hydroxy-3-nitrobenzaldehyde
Molecular FormulaC7H5NO4
InChI KeyNUGOTBXFVWXVTE-UHFFFAOYSA-N
SMILESOC1=C(C=O)C=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormPowder
Assay (GC)≥97.5%
Melting Point (clear melt)107.0-113.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only