Thermo Scientific Chemicals

1-Iodo-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B23659.06
5 g, Each
Thermo Scientific Chemicals

1-Iodo-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B23659.06
5 g, Each
Quantity
Catalog number: B23659.06
also known as B23659-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
709-49-9
IUPAC Name
1-iodo-2,4-dinitrobenzene
Molecular Formula
C6H3IN2O4
InChI Key
FXMKXMJLXRTQSW-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(I)C(=C1)[N+]([O-])=O
Melting Point (clear melt)
85.0-91.0?C
Appearance (Color)
Yellow to orange to brown
Form
Powder
Assay (GC)
≥97.5%

Description

It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text