Thermo Scientific Chemicals

Iminodiacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: B23660.36
500 g, Each
Thermo Scientific Chemicals

Iminodiacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: B23660.36
500 g, Each
Quantity
Catalog number: B23660.36
also known as B23660-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-87-5
IUPAC Name
2-[(cyanomethyl)amino]acetonitrile
Molecular Formula
C4H5N3
InChI Key
BSRDNMMLQYNQQD-UHFFFAOYSA-N
SMILES
N#CCNCC#N
Appearance (Color)
White to cream to brown
Form
Crystals or crystalline powder or powder
Assay (Non-aqueous acid-base Titration)
≥90.0 to ≤110.0%
Melting Point (clear melt)
72-82°C

Description

Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

Solubility
Soluble in water, acetone and hot methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text