Thermo Scientific Chemicals

2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23662.09
10 g, Each
Thermo Scientific Chemicals

2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B23662.09
10 g, Each
Quantity
Catalog number: B23662.09
also known as B23662-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7148-74-5
IUPAC Name
2-bromopropanoyl chloride
Molecular Formula
C3H4BrClO
InChI Key
OZGMODDEIHYPRY-UHFFFAOYNA-N
SMILES
CC(Br)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow to yellow to orange
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Refractive Index
1.4760-1.4800 @ 20?C
Form
Liquid

Description

2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.

Solubility
Reacts with water.

Notes
Protect from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep away from ignition sources. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text