2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.
Solubility
Reacts with water.
Notes
Protect from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep away from ignition sources. Protect from electrostatic charges.
RUO – Research Use Only
General References:
- Katja Jankova, et al. Synthesis of poly(styrene-b-isobutylene-b-styrene) triblock copolymer by ATRP.Polym. Bull.,1998,41(6), 639-644.
- Su-Sun. Wang. Solid phase synthesis of protected peptides via photolytic cleavage of the .alpha.-methylphenacyl ester anchoring linkage.J. Org. Chem.,1976,41(20), 3258-3261.