n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number B23673.AE
also known as B23673-AE
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
Add to cart
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH, Thermo Scientific Chemicals
Catalog numberB23673.AE
Price (USD)/ Each
30.65
Online exclusive
34.00 
Save 3.35 (10%)
-
Add to cart
Chemical Identifiers
CAS111-34-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Solution Test10% in diethyl ether: clear
Assay (GC)≥97.5%
Refractive Index1.3990-1.4030 @ 20?C
View more
Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 3 g/L (20°C). Soluble in chloroform and methanol.

Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.
RUO – Research Use Only

General References:

  1. Franz Karl Fleischmann, et al. Cycloaddition reaction between tretracyanoethylene and n-butyl vinyl ether: The solvent dependence of the volume of activation.Tetrahedron Lett.,1973,14(39), 3773-3776.
  2. Chang-Min Xing, et al. Surface Photo grafting Polymerization of Binary Monomers Maleic Anhydride and n-Butyl Vinyl Ether on Polypropylene Film I. Effects of Principal Factors.Polym. J.,2002,34801-808.