Thermo Scientific Chemicals

Benzo[b]furan-2-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23676.03
1 g, Each
Thermo Scientific Chemicals

Benzo[b]furan-2-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23676.03
1 g, Each
Quantity
Catalog number: B23676.03
also known as B23676-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98437-24-2
IUPAC Name
(1-benzofuran-2-yl)boronic acid
Molecular Formula
C8H7BO3
InChI Key
PKRRNTJIHGOMRC-UHFFFAOYSA-N
SMILES
OB(O)C1=CC2=CC=CC=C2O1
Form
Powder
Appearance (Color)
Pale yellow to pale brown
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

Solubility
Soluble in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text