It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.
Solubility
Soluble in water.
Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
RUO – Research Use Only
Yu Mi Chang, et al. Petasis reaction of activated quinoline and isoquinoline with various boronic acids.Tetrahedron Lett.,2005,46(17), 3053-3056.
Jason R. Schmink, et al. Facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.Tetrahedron Lett.,2013,54(1), 15-20.