4-Ethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Ethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B23683.06
also known as B23683-06
Price (USD)/ Each
78.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.40
Add to cart
4-Ethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberB23683.06
Price (USD)/ Each
78.40
-
Add to cart
Chemical Identifiers
CAS22237-13-4
IUPAC Name(4-ethoxyphenyl)boronic acid
Molecular FormulaC8H11BO3
InChI KeyWRQNDLDUNQMTCL-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream
Assay (HPLC)≥97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only