4-Chlorobenzhydrol, 98%, Thermo Scientific Chemicals
4-Chlorobenzhydrol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzhydrol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23691.06
also known as B23691-06
Price (USD)/ Each
43.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
43.70
Add to cart
4-Chlorobenzhydrol, 98%, Thermo Scientific Chemicals
Catalog numberB23691.06
Price (USD)/ Each
43.70
-
Add to cart
Chemical Identifiers
CAS119-56-2
IUPAC Name(4-chlorophenyl)(phenyl)methanol
Molecular FormulaC13H11ClO
InChI KeyAJYOOHCNOXWTKJ-UHFFFAOYNA-N
SMILESOC(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)58.0-64.0?C
Assay (GC)≥97.5%
FormPowder or lumps
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only