2-Methoxy-5-sulfamoylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Methoxy-5-sulfamoylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxy-5-sulfamoylbenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B23698.09
also known as B23698-09
Price (USD)/ Each
53.30
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
53.30
Add to cart
2-Methoxy-5-sulfamoylbenzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberB23698.09
Price (USD)/ Each
53.30
-
Add to cart
Chemical Identifiers
CAS22117-85-7
IUPAC Name2-methoxy-5-sulfamoylbenzoate
Molecular FormulaC8H8NO5S
InChI KeySQAILWDRVDGLGY-UHFFFAOYSA-M
SMILESCOC1=CC=C(C=C1C([O-])=O)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
Appearance (Color)White
Assay (HPLC)≥96.0%
Melting Point (clear melt)218.0-227.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only