4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B23709.06
also known as B23709-06
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
Add to cart
4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberB23709.06
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2156-04-9
IUPAC Name(4-ethenylphenyl)boronic acid
Molecular FormulaC8H9BO2
InChI KeyQWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
View more
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Vancoillie, G.; Pelz, S.; Holder, E.; Hoogenboom, R. Direct nitroxide mediated (co)polymerization of 4-vinylphenylboronic acid as route towards sugar sensors. Polym. Chem. 2012, 3 (7), 1726-1729.
  2. Maji, S.; Vancoillie, G.; Voorhaar, L.; Zhang, Q.; Hoogenboom, R. RAFT Polymerization of 4-Vinylphenylboronic Acid as the Basis for Micellar Sugar Sensors. Macromol. Rapid Commun. 2014, 35 (2), 214-220.