Thermo Scientific Chemicals

4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23709.14
25 g, Each
Thermo Scientific Chemicals

4-Vinylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23709.14
25 g, Each
Quantity
Catalog number: B23709.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2156-04-9
IUPAC Name
(4-ethenylphenyl)boronic acid
Molecular Formula
C8H9BO2
InChI Key
QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(C=C)C=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Proton NMR
Conforms to structure

Description

4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text