N-Benzylformamide, 99%, Thermo Scientific Chemicals
N-Benzylformamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylformamide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B23722.22
also known as B23722-22
Price (USD)/ Each
568.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
568.00
Add to cart
N-Benzylformamide, 99%, Thermo Scientific Chemicals
Catalog numberB23722.22
Price (USD)/ Each
568.00
-
Add to cart
Chemical Identifiers
CAS6343-54-0
IUPAC NameN-benzylformamide
Molecular FormulaC8H9NO
InChI KeyIIBOGKHTXBPGEI-UHFFFAOYSA-N
SMILESO=CNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormPowder
Assay (GC)≥98.5%
Melting Point (clear melt)58.0-64.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only