Thermo Scientific Chemicals

(3-Mercaptopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: B23726.14
25 g, Each
Thermo Scientific Chemicals

(3-Mercaptopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: B23726.14
25 g, Each
Quantity
Catalog number: B23726.14
also known as B23726-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4420-74-0
IUPAC Name
3-(trimethoxysilyl)propane-1-thiol
Molecular Formula
C6H16O3SSi
InChI Key
UUEWCQRISZBELL-UHFFFAOYSA-N
SMILES
CO[Si](CCCS)(OC)OC
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms (UK Sourced Material)
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥94.0%

Description

(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

Solubility
Miscible with methanol, ethanol, isopropanol, acetone, benzene, toluene and xylene.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, alcohols and water. Decomposes in water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text