Thermo Scientific Chemicals

1-Pyrrolidinecarbodithioic acid ammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: B23731.22
100 g, Each
Thermo Scientific Chemicals

1-Pyrrolidinecarbodithioic acid ammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: B23731.22
100 g, Each
Quantity
Catalog number: B23731.22
also known as B23731-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5108-96-3
IUPAC Name
pyrrolidine-1-carbodithioic acid amine
Molecular Formula
C5H12N2S2
InChI Key
MDDIUTVUBYEEEM-UHFFFAOYSA-N
SMILES
N.SC(=S)N1CCCC1
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

Solubility
Soluble in water.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text