2,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
2,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B23740.06
also known as B23740-06
Price (USD)/ Each
90.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
90.90
Add to cart
2,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberB23740.06
Price (USD)/ Each
90.90
-
Add to cart
Chemical Identifiers
CAS85199-06-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5%
FormPowder
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only