Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B23744.14
also known as B23744-14
Price (USD)/ Each
81.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
81.70
Add to cart
Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals
Catalog numberB23744.14
Price (USD)/ Each
81.70
-
Add to cart
Chemical Identifiers
CAS1115-30-6
IUPAC Name1,4-diethyl 2-acetylbutanedioate
Molecular FormulaC10H16O5
InChI KeyDVSDDICSXBCMQJ-UHFFFAOYNA-N
SMILESCCOC(=O)CC(C(C)=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Refractive Index1.4325-1.4375 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only