Thermo Scientific Chemicals

Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals

Catalog number: B23744.22
100 g, Each
Thermo Scientific Chemicals

Diethyl acetylsuccinate, 97+%, Thermo Scientific Chemicals

Catalog number: B23744.22
100 g, Each
Quantity
Catalog number: B23744.22
also known as B23744-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1115-30-6
IUPAC Name
1,4-diethyl 2-acetylbutanedioate
Molecular Formula
C10H16O5
InChI Key
DVSDDICSXBCMQJ-UHFFFAOYNA-N
SMILES
CCOC(=O)CC(C(C)=O)C(=O)OCC
Assay (GC)
≥97.0%
Refractive Index
1.4325-1.4375 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text