Thermo Scientific Chemicals

Tetrakis(dimethylsiloxy)silane, 97%, Thermo Scientific Chemicals

Catalog number: B23754.22
100 g, Each
Thermo Scientific Chemicals

Tetrakis(dimethylsiloxy)silane, 97%, Thermo Scientific Chemicals

Catalog number: B23754.22
100 g, Each
Quantity
Catalog number: B23754.22
also known as B23754-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17082-47-2
Molecular Formula
C8H24O4Si5
InChI Key
UOUILILVWRHZSH-UHFFFAOYSA-N
SMILES
C[Si](C)O[Si](O[Si](C)C)(O[Si](C)C)O[Si](C)C
Molecular Weight (g/mol)
324.70
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.3850-1.3890 @ 20?C

Description

It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text