Thermo Scientific Chemicals

Hexanoic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B23756.30
250 g, Each
Thermo Scientific Chemicals

Hexanoic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B23756.30
250 g, Each
Quantity
Catalog number: B23756.30
also known as B23756-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2051-49-2
IUPAC Name
hexanoyl hexanoate
Molecular Formula
C12H22O3
InChI Key
PKHMTIRCAFTBDS-UHFFFAOYSA-N
SMILES
CCCCCC(=O)OC(=O)CCCCC
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4265-1.4305 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text