Thermo Scientific Chemicals

2-Amino-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B23773.22
100 g, Each
Thermo Scientific Chemicals

2-Amino-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B23773.22
100 g, Each
Quantity
Catalog number: B23773.22
also known as B23773-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1775-95-7
Appearance (Color)
Yellow to brown
Form
Crystalline powder
Assay (GC)
>97.5%

Description

It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.

Solubility
Soluble in chloroform, dichloromethane, dimethyl sulfoxide, ethyl acetate and methanol. Soluble in water 3 mg/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text