2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, amines, alcohols, active metals, water/ moisture.
RUO – Research Use Only
Eugene H. Man; Frederic W. Swamer; Charles R. Hauser. The Claisen Acylation of Methyl Ketones with Branched Chain Aliphatic Esters. J. Am. Chem. Soc. 1951, 73 (3), 901-903.
Loren M. Long; Henry R. Henze. Synthesis of Ketone Derivatives of Biphenyl by the Friedel-Crafts Reaction. J. Am. Chem. Soc. 1941, 63 (7), 1939-1940.