Thermo Scientific Chemicals

1,2,4-Tribromobenzene, 95%, Thermo Scientific Chemicals

Catalog number: B23778.14
25 g, Each
Thermo Scientific Chemicals

1,2,4-Tribromobenzene, 95%, Thermo Scientific Chemicals

Catalog number: B23778.14
25 g, Each
Quantity
Catalog number: B23778.14
also known as B23778-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-54-3
IUPAC Name
1,2,4-tribromobenzene
Molecular Formula
C6H3Br3
InChI Key
FWAJPSIPOULHHH-UHFFFAOYSA-N
SMILES
BrC1=CC=C(Br)C(Br)=C1
Appearance (Color)
White to grey or cream or pale yellow to pale brown
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥94.0%
Melting Point (clear melt)
40-44?C

Description

1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text