It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from heavy metal powders and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
RUO – Research Use Only
Benjamin S Flavel, et al. Grafting of poly(ethylene glycol) on click chemistry modified Si(100) surfaces.Langmuir.,2013,72(1), 8355-8362.
Néstor M Carballeira, et al. 2,6-hexadecadiynoic acid and 2,6-nonadecadiynoic acid: novel synthesized acetylenic fatty acids as potent antifungal agents.Lipids.,2006,41(5), 507-11.