Thermo Scientific Chemicals

Tribenzylamine, 99+%, Thermo Scientific Chemicals

Catalog number: B23797.14
25 g, Each
Thermo Scientific Chemicals

Tribenzylamine, 99+%, Thermo Scientific Chemicals

Catalog number: B23797.14
25 g, Each
Quantity
Catalog number: B23797.14
also known as B23797-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-40-6
IUPAC Name
tribenzylamine
Molecular Formula
C21H21N
InChI Key
MXHTZQSKTCCMFG-UHFFFAOYSA-N
SMILES
C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Identification (FTIR)
Conforms
Assay (GC)
≥99.0%
Water Content (Karl Fischer Titration)
≤0.5%
Form
Crystals or powder or crystalline powder

Description

Has been used as a base in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text