Thermo Scientific Chemicals

1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog number: B23799.22
100 g, Each
Thermo Scientific Chemicals

1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog number: B23799.22
100 g, Each
Quantity
Catalog number: B23799.22
also known as B23799-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-94-5
IUPAC Name
1-methylpyrrolidine
Molecular Formula
C5H11N
InChI Key
AVFZOVWCLRSYKC-UHFFFAOYSA-N
SMILES
CN1CCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4225-1.4265 @ 20?C

Description

1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text