Thermo Scientific Chemicals

Diisopropyl phosphite, 98%, Thermo Scientific Chemicals

Catalog number: B23800.14
25 g, Each
Thermo Scientific Chemicals

Diisopropyl phosphite, 98%, Thermo Scientific Chemicals

Catalog number: B23800.14
25 g, Each
Quantity
Catalog number: B23800.14
also known as B23800-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1809-20-7
IUPAC Name
oxobis(propan-2-yloxy)-λ⁵-phosphanylium
Molecular Formula
C6H14O3P
InChI Key
BJLZAAWLLPMZQR-UHFFFAOYSA-N
SMILES
CC(C)O[P+](=O)OC(C)C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4070-1.4110 @ 20?C

Description

Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

Solubility
Soluble in water, chloroform.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text