3-Fluoro-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Fluoro-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23827.03
also known as B23827-03
Price (USD)/ Each
53.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
53.50
Add to cart
3-Fluoro-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB23827.03
Price (USD)/ Each
53.50
-
Add to cart
Chemical Identifiers
CAS351-54-2
IUPAC Name3-fluoro-4-methoxybenzaldehyde
Molecular FormulaC8H7FO2
InChI KeySOQCZBSZZLWDGU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)28.0-35.0?C
Free acid (titration)≤1%
Appearance (Color)Cream to yellow
FormFused solid or clear liquid as melt
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only