Thermo Scientific Chemicals

Heptanal, 97%, Thermo Scientific Chemicals

Catalog number: B23830.AP
500 mL, Each
Thermo Scientific Chemicals

Heptanal, 97%, Thermo Scientific Chemicals

Catalog number: B23830.AP
500 mL, Each
Quantity
Catalog number: B23830.AP
also known as B23830-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-71-7
IUPAC Name
heptanal
Molecular Formula
C7H14O
InChI Key
FXHGMKSSBGDXIY-UHFFFAOYSA-N
SMILES
CCCCCCC=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Free acid (titration)
≤2.0%
Identification (FTIR)
Conforms
Refractive Index
1.4100-1.4150 @ 20?C

Description

Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

Solubility
Soluble in water 1.3 g/L.

Notes
Hygroscopic. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text