Thermo Scientific Chemicals

1-Adamantanemethanol, 98%, Thermo Scientific Chemicals

Catalog number: B23831.06
5 g, Each
Thermo Scientific Chemicals

1-Adamantanemethanol, 98%, Thermo Scientific Chemicals

Catalog number: B23831.06
5 g, Each
Quantity
Catalog number: B23831.06
also known as B23831-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
770-71-8
IUPAC Name
(adamantan-1-yl)methanol
Molecular Formula
C11H18O
InChI Key
MDVGOOIANLZFCP-UHFFFAOYSA-N
SMILES
OCC12CC3CC(CC(C3)C1)C2
Appearance (Color)
White to pale cream
Assay (GC)
>97.5%
Form
Crystals, Crystalline powder and/or Lumps

Description

1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text