3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23833.03
also known as B23833-03
Price (USD)/ Each
77.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
77.30
Add to cart
3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberB23833.03
Price (USD)/ Each
77.30
-
Add to cart
Chemical Identifiers
CAS78887-39-5
IUPAC Name(3-acetamidophenyl)boronic acid
Molecular FormulaC8H10BNO3
InChI KeyIBTSWKLSEOGJGJ-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
Appearance (Color)Pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only