3-Cyanobenzoic acid, 98+%, Thermo Scientific Chemicals
3-Cyanobenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Cyanobenzoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B23836.22
also known as B23836-22
Price (USD)/ Each
577.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
577.00
Add to cart
3-Cyanobenzoic acid, 98+%, Thermo Scientific Chemicals
Catalog numberB23836.22
Price (USD)/ Each
577.00
-
Add to cart
Chemical Identifiers
CAS1877-72-1
IUPAC Name3-cyanobenzoic acid
Molecular FormulaC8H5NO2
InChI KeyGYLKKXHEIIFTJH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale gray
Melting Point (clear melt)218.0-224.0?C
FormPowder
Assay (HPLC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only