Thermo Scientific Chemicals

Cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B23837.14
25 g, Each
Thermo Scientific Chemicals

Cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B23837.14
25 g, Each
Quantity
Catalog number: B23837.14
also known as B23837-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2043-61-0
IUPAC Name
cyclohexanecarbaldehyde
Molecular Formula
C7H12O
InChI Key
KVFDZFBHBWTVID-UHFFFAOYSA-N
SMILES
O=CC1CCCCC1
Identification (FTIR)
Conforms
Refractive Index
1.4480-1.4540 @ 20°C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%

Description

Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with ethyl diazoacetate catalyzed by basic, nanoparticulate magnesium oxide leading to α-diazo-β-hydroxyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with eth
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text