4-Chlorothiophenol, 97%, Thermo Scientific Chemicals
4-Chlorothiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorothiophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23838.14
also known as B23838-14
Price (USD)/ Each
28.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
28.70
Add to cart
4-Chlorothiophenol, 97%, Thermo Scientific Chemicals
Catalog numberB23838.14
Price (USD)/ Each
28.70
-
Add to cart
Chemical Identifiers
CAS106-54-7
IUPAC Name4-chlorobenzene-1-thiol
Molecular FormulaC6H5ClS
InChI KeyVZXOZSQDJJNBRC-UHFFFAOYSA-N
SMILESSC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps or fused solid
Melting Point (clear melt)48.0-57.0°C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only