Thermo Scientific Chemicals

Ergosterol, 96%, may cont. up to ca 6% water, Thermo Scientific Chemicals

Catalog number: B23840.14
25 g, Each
Thermo Scientific Chemicals

Ergosterol, 96%, may cont. up to ca 6% water, Thermo Scientific Chemicals

Catalog number: B23840.14
25 g, Each
Quantity
Catalog number: B23840.14
also known as B23840-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-87-4
IUPAC Name
(1R,3aR,7S,9aR,9bS,11aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular Formula
C28H44O
InChI Key
DNVPQKQSNYMLRS-APGDWVJJSA-N
SMILES
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Optical Rotation
-120? ? 15? (c=1 in Chloroform)
Form
Crystals or powder or crystalline powder or needles
Appearance (Color)
White to cream
Assay (HPLC)
≥95.0%

Description

Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

Solubility
Soluble in water (<1mg/mL at 25°C), chloroform (50 mg/ml), DMSO (<1mg/mL at 25°C), ethanol (<1mg/mL at 25°C), and ether (14.29 mg/ml).

Notes
Light sensitive. Store away from air, light and oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text