Thermo Scientific Chemicals

Furan-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23842.03
1 g, Each
Thermo Scientific Chemicals

Furan-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23842.03
1 g, Each
Quantity
Catalog number: B23842.03
also known as B23842-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13331-23-2
IUPAC Name
(furan-2-yl)boronic acid
Molecular Formula
C4H5BO3
InChI Key
PZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CO1
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%
Proton NMR
Conforms to structure
Assay (HPLC)
≥96.0%
Appearance (Color)
Pale cream to cream

Description

It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text