Thermo Scientific Chemicals

1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals

Catalog number: B23854.30
250 g, Each
Thermo Scientific Chemicals

1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals

Catalog number: B23854.30
250 g, Each
Quantity
Catalog number: B23854.30
also known as B23854-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-77-3
IUPAC Name
1-bromo-2-methylpropane
Molecular Formula
C4H9Br
InChI Key
HLVFKOKELQSXIQ-UHFFFAOYSA-N
SMILES
CC(C)CBr
Identification (FTIR)
Conforms
Refractive Index
1.4340-1.4380 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%

Description

1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

Solubility
Slightly soluble in water. Miscible with alcohol, ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text