Thermo Scientific Chemicals

2-Phenylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B23858.22
100 g, Each
Thermo Scientific Chemicals

2-Phenylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B23858.22
100 g, Each
Quantity
Catalog number: B23858.22
also known as B23858-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
670-96-2
IUPAC Name
2-phenyl-1H-imidazole
Molecular Formula
C9H8N2
InChI Key
ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
SMILES
N1C=CN=C1C1=CC=CC=C1
Assay (GC)
≥97.5%
Appearance (Color)
Pale yellow
Melting Point (clear melt)
142.0-152.0°C
Form
Crystals or powder or crystalline powder

Description

2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

Solubility
Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text