Thermo Scientific Chemicals

1,3,5-Tribromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B23860.14
25 g, Each
Thermo Scientific Chemicals

1,3,5-Tribromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B23860.14
25 g, Each
Quantity
Catalog number: B23860.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-39-1
IUPAC Name
1,3,5-tribromobenzene
Molecular Formula
C6H3Br3
InChI Key
YWDUZLFWHVQCHY-UHFFFAOYSA-N
SMILES
BrC1=CC(Br)=CC(Br)=C1
Appearance (Color)
White to cream to brown
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Melting Point
117.5-123.5?C
Form
Crystals or powder or crystalline powder

Description

1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text