Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: B23867.22
100 g, Each
Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: B23867.22
100 g, Each
Quantity
Catalog number: B23867.22
also known as B23867-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-54-2
IUPAC Name
N1-phenylbenzene-1,4-diamine
Molecular Formula
C12H12N2
InChI Key
ATGUVEKSASEFFO-UHFFFAOYSA-N
SMILES
NC1=CC=C(NC2=CC=CC=C2)C=C1
Form
Crystals or powder or crystalline powder and/or chunks or flakes
Appearance (Color)
Dark brown or dark purple to dark gray
Assay (GC)
≥97.5%
Melting Point (clear melt)
70.0-77.0°C

Description

p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.

Solubility
Soluble in water at 20°C 0.6g/L. Soluble in (10 mg/mL) ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text