Thermo Scientific Chemicals

1,8-Diaminooctane, 98%, Thermo Scientific Chemicals

Catalog number: B23885.22
100 g, Each
Thermo Scientific Chemicals

1,8-Diaminooctane, 98%, Thermo Scientific Chemicals

Catalog number: B23885.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B23885.22
also known as B23885-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
373-44-4
IUPAC Name
octane-1,8-bis(aminium) dichloride
Molecular Formula
C8H22Cl2N2
InChI Key
ZFLWZOGXFQNIMT-UHFFFAOYSA-N
SMILES
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
Assay (GC)
≥97.5%
Melting Point (clear melt)
49.0-56.0?C
Form
Crystals or powder or crystalline powder or chunks or fused solid
Appearance (Color)
White to cream to pale yellow
Identification (FTIR)
Conforms

Description

1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water 575g/L.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, carbon dioxide, air, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text