Thermo Scientific Chemicals

Pyridine-2-carboxaldoxime methochloride, 97%, Thermo Scientific Chemicals

Catalog number: B23890.14
25 g, Each
Thermo Scientific Chemicals

Pyridine-2-carboxaldoxime methochloride, 97%, Thermo Scientific Chemicals

Catalog number: B23890.14
25 g, Each
Quantity
Catalog number: B23890.14
also known as B23890-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-15-0
IUPAC Name
(2E)-1-methyl-2-(nitrosomethylidene)-1,2-dihydropyridine hydrochloride
Molecular Formula
C7H9ClN2O
InChI Key
HIGSLXSBYYMVKI-UHDJGPCESA-N
SMILES
Cl.CN1C=CC=C\C1=C/N=O
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text