1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals
1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B23904.18
also known as B23904-18
Price (USD)/ Each
37.50
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
37.50
Add to cart
1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals
Catalog numberB23904.18
Price (USD)/ Each
37.50
-
Add to cart
Chemical Identifiers
CAS99-62-7
IUPAC Name1,3-bis(propan-2-yl)benzene
Molecular FormulaC12H18
InChI KeyUNEATYXSUBPPKP-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=CC=C1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥95.0%
Comment1.4870-1.4905 @ 20°C
Identification (FTIR)Conforms
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

Solubility
Soluble in water 4.33 mg/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A. Corma; V. Fornes; S. B. Pergher; Th. L. M. Maesen & J. G. Buglass. Delaminated zeolite precursors as selective acidic catalysts.Nature.1998, 396 353-356.
  2. Sharif F. Zaman; Kevin F. Loughlin and Sulaiman S. Al-Khattaf. Kinetics of Desorption of 1,3-Diisopropylbenzene and 1,3,5-Triisopropylbenzene. 1. Diffusion in Y-Zeolite Crystals by the Zero-Length-Column Method. Ind. Eng. Chem. Res. 2005, 44 (7), 2027-2035.