Thermo Scientific Chemicals

1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals

Catalog number: B23904.30
250 g, Each
Thermo Scientific Chemicals

1,3-Diisopropylbenzene, 96%, Thermo Scientific Chemicals

Catalog number: B23904.30
250 g, Each
Quantity
Catalog number: B23904.30
also known as B23904-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-62-7
IUPAC Name
1,3-bis(propan-2-yl)benzene
Molecular Formula
C12H18
InChI Key
UNEATYXSUBPPKP-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=CC=C1)C(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%
Comment
1.4870-1.4905 @ 20°C
Identification (FTIR)
Conforms

Description

1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

Solubility
Soluble in water 4.33 mg/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text