Thermo Scientific Chemicals

Trichloroisocyanuric acid, 90+%, Thermo Scientific Chemicals

Catalog number: B23906.0E
2500 g, Each
Thermo Scientific Chemicals

Trichloroisocyanuric acid, 90+%, Thermo Scientific Chemicals

Catalog number: B23906.0E
2500 g, Each
Quantity
Catalog number: B23906.0E
also known as B23906-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-90-1
IUPAC Name
trichloro-1,3,5-triazinane-2,4,6-trione
Molecular Formula
C3Cl3N3O3
InChI Key
YRIZYWQGELRKNT-UHFFFAOYSA-N
SMILES
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
Appearance (Color)
White to pale cream
Form
Granules or powder
Assay (Iodometric Titration)
≥90% to ≤110% (non-U.S. specification)
Assay from Suppliers CofA
≥90% to ≤110% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.

Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).

Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text