Thermo Scientific Chemicals

Methyl nicotinate, 99%, Thermo Scientific Chemicals

Catalog number: B23908.30
250 g, Each
Thermo Scientific Chemicals

Methyl nicotinate, 99%, Thermo Scientific Chemicals

Catalog number: B23908.30
250 g, Each
Quantity
Catalog number: B23908.30
also known as B23908-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-60-7
IUPAC Name
methyl pyridine-3-carboxylate
Molecular Formula
C7H7NO2
InChI Key
YNBADRVTZLEFNH-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CN=C1
Appearance (Color)
White to pale cream to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
37.0-44.0?C

Description

Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

Solubility
Soluble in water at 20°C 48 g/L. Soluble in alcohol, chloroform and ethyl acetate. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text