4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B23915.14
also known as B23915-14
Price (USD)/ Each
82.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
82.90
Add to cart
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
Catalog numberB23915.14
Price (USD)/ Each
82.90
-
Add to cart
Chemical Identifiers
CAS2481-94-9
IUPAC NameN,N-diethyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC16H19N3
InChI KeySJJISKLXUJVZOA-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.0%
Assay (GC)≥97.5%
Appearance (Color)Yellow to orange to brown
Melting Point (clear melt)91.0-98.0°C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only